The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193380.1 | NP_001180309.1 | 7 - 8 | 835 | 70 |
NM_153480.1 | NP_705613.1 | 7 - 8 | 835 | 70 | |
NM_153481.1 | NP_705614.1 | 5 - 6 | 699 | 70 | |
NM_153483.2 | NP_705616.2 | 8 - 9 | 854 | 70 | |
NR_104198.1 | 6 - 7 | 819 | 70 | ||
XM_006712975.1 | XP_006713038.1 | 6 - 7 | 661 | 70 | |
XM_006712976.1 | XP_006713039.1 | 6 - 7 | 799 | 70 | |
XM_011533361.2 | XP_011531663.1 | 7 - 8 | 751 | 70 | |
XM_011533362.2 | XP_011531664.1 | 7 - 8 | 770 | 70 | |
XM_011533363.2 | XP_011531665.1 | 7 - 8 | 770 | 70 | |
XM_011533364.2 | XP_011531666.1 | 7 - 8 | 770 | 70 | |
GenBank mRNA | AF458069.1 | - | 7 - 8 | 835 | 70 |
AK074238.1 | - | 7 - 8 | 815 | 70 | |
AK096793.1 | - | 6 - 7 | 797 | 70 | |
AK096801.1 | - | 5 - 6 | 679 | 70 | |
AK297697.1 | - | 7 - 8 | 801 | 70 | |
AK314476.1 | - | 7 - 8 | 820 | 70 | |
AY358451.1 | - | 7 - 8 | 835 | 70 | |
BC035478.1 | - | 8 - 9 | 847 | 70 | |
BC137077.1 | - | 8 - 9 | 797 | 70 | |
BC137078.1 | - | 8 - 9 | 797 | 70 |