The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_058216.2 | NP_478123.1 | 6 - 7 | 61 | |
NR_103872.1 | 5 - 6 | 61 | |||
XM_006722001.3 | XP_006722064.1 | 6 - 7 | 61 | ||
XM_006722004.3 | XP_006722067.1 | 6 - 7 | 61 | ||
XM_006722005.3 | XP_006722068.1 | 6 - 7 | 61 | ||
XM_011525092.2 | XP_011523394.1 | 6 - 7 | 61 | ||
XM_011525093.2 | XP_011523395.1 | 6 - 7 | 61 | ||
XM_011525094.2 | XP_011523396.1 | 6 - 7 | 61 | ||
XM_017024914.1 | XP_016880403.1 | 6 - 7 | 61 | ||
XM_017024915.1 | XP_016880404.1 | 6 - 7 | 61 | ||
XM_017024916.1 | XP_016880405.1 | 7 - 8 | 61 | ||
XM_017024917.1 | XP_016880406.1 | 6 - 7 | 61 | ||
XM_017024918.1 | XP_016880407.1 | 6 - 7 | 61 | ||
GenBank mRNA | AF029669.1 | - | 6 - 7 | 61 | |
AK303363.1 | - | 6 - 7 | 61 | ||
BC000667.2 | - | 5 - 6 | 61 | ||
BC107753.1 | - | 6 - 7 | 61 | ||
CB243543.1 | - | 3 - 4 | 358 | 61 |