Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLE6
Entrez Gene ID
79816
Gene Name
transducin like enhancer of split 6
Gene Aliases
GRG6, PREMBL
Chromosome Location
Chr.19: 2977410 - 2995184 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143986.1 NP_001137458.1 7 - 8 656 65
NM_024760.2 NP_079036.1 6 - 7 578 65
XM_005259645.2 XP_005259702.1 7 - 8 665 65
XM_011528300.2 XP_011526602.1 7 - 8 641 65
XM_011528301.1 XP_011526603.1 3 - 4 368 65
GenBank mRNA AK024071.1 - 7 - 8 631 65
BC007215.1 - 1 - 2 264 65
BC013986.2 - 6 - 7 536 65
BC020206.1 - 7 - 8 613 65

View Full Product Details