The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164391.1 | NP_001157863.1 | 9 - 10 | 1005 | 155 |
NM_001164392.1 | NP_001157864.1 | 8 - 9 | 1085 | 155 | |
NM_001164393.1 | NP_001157865.1 | 6 - 7 | 804 | 155 | |
NM_007358.3 | NP_031384.1 | 8 - 9 | 1085 | 155 | |
XM_011541016.1 | XP_011539318.1 | 8 - 9 | 1041 | 155 | |
XM_011541017.2 | XP_011539319.1 | 8 - 9 | 1101 | 155 | |
XM_017000676.1 | XP_016856165.1 | 8 - 9 | 940 | 155 | |
XM_017000677.1 | XP_016856166.1 | 8 - 9 | 1041 | 155 | |
XM_017000678.1 | XP_016856167.1 | 9 - 10 | 994 | 155 | |
GenBank mRNA | AF072814.1 | - | 7 - 8 | 783 | 155 |
AJ010014.1 | - | 8 - 9 | 1036 | 155 | |
AK290318.1 | - | 8 - 9 | 1037 | 155 | |
AK302776.1 | - | 9 - 10 | 944 | 155 | |
AK302900.1 | - | 6 - 7 | 756 | 155 | |
BC001897.1 | - | 8 - 9 | 1004 | 155 | |
BC010013.2 | - | 8 - 9 | 1009 | 155 |