Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AKNA
Entrez Gene ID
80709
Gene Name
AT-hook transcription factor
Gene Aliases
-
Chromosome Location
Chr.9: 114334149 - 114398498 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317950.1 NP_001304879.1 20 - 21 4091 85
NM_001317952.1 NP_001304881.1 19 - 20 3649 85
NM_030767.5 NP_110394.3 20 - 21 4036 85
XM_005252244.2 XP_005252301.1 22 - 23 4208 85
XM_005252245.1 XP_005252302.1 21 - 22 4066 85
XM_005252247.4 XP_005252304.1 20 - 21 4087 85
XM_005252248.2 XP_005252305.1 19 - 20 4108 85
XM_006717294.1 XP_006717357.1 20 - 21 4020 85
XM_011519063.2 XP_011517365.1 19 - 20 3699 85
XM_011519065.2 XP_011517367.2 20 - 21 3679 85
XM_017015172.1 XP_016870661.1 21 - 22 3806 85
GenBank mRNA AF286341.1 - 10 - 11 1590 85
AK024431.1 - 18 - 19 2984 85
AK074040.1 - 19 - 20 3079 85
AK131082.2 - 20 - 21 4091 85
AK160382.1 - 20 - 21 4093 85
AL713755.1 - 3 - 4 2525 85
AL833220.1 - 17 - 18 2513 85
AY703039.1 - 21 - 22 3691 85
AY703040.1 - 19 - 20 3649 85
AY703043.1 - 19 - 20 4009 85
AY703044.1 - 18 - 19 3703 85
AY703045.1 - 11 - 12 1782 85
AY703046.1 - 7 - 8 908 85
BC042202.1 - 6 - 7 766 85
BC055285.1 - 20 - 21 4036 85

View Full Product Details