The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317950.1 | NP_001304879.1 | 9 - 10 | 2338 | 53 |
NM_001317952.1 | NP_001304881.1 | 8 - 9 | 1896 | 53 | |
NM_030767.5 | NP_110394.3 | 9 - 10 | 2283 | 53 | |
XM_005252244.2 | XP_005252301.1 | 11 - 12 | 2455 | 53 | |
XM_005252245.1 | XP_005252302.1 | 10 - 11 | 2313 | 53 | |
XM_005252247.4 | XP_005252304.1 | 9 - 10 | 2334 | 53 | |
XM_005252248.2 | XP_005252305.1 | 9 - 10 | 2451 | 53 | |
XM_006717294.1 | XP_006717357.1 | 9 - 10 | 2267 | 53 | |
XM_006717295.2 | XP_006717358.1 | 9 - 10 | 2451 | 53 | |
XM_011519063.2 | XP_011517365.1 | 8 - 9 | 1946 | 53 | |
XM_011519064.2 | XP_011517366.1 | 9 - 10 | 2451 | 53 | |
XM_011519065.2 | XP_011517367.2 | 9 - 10 | 1926 | 53 | |
XM_011519066.2 | XP_011517368.1 | 9 - 10 | 2451 | 53 | |
XM_017015172.1 | XP_016870661.1 | 10 - 11 | 2053 | 53 | |
GenBank mRNA | AB075848.1 | - | 9 - 10 | 2456 | 53 |
AK024431.1 | - | 7 - 8 | 1231 | 53 | |
AK074040.1 | - | 8 - 9 | 1326 | 53 | |
AK131082.2 | - | 9 - 10 | 2338 | 53 | |
AK160382.1 | - | 9 - 10 | 2338 | 53 | |
AL833220.1 | - | 6 - 7 | 760 | 53 | |
AY703039.1 | - | 10 - 11 | 1938 | 53 | |
AY703040.1 | - | 8 - 9 | 1896 | 53 | |
AY703041.1 | - | 10 - 11 | 1652 | 53 | |
AY703042.1 | - | 8 - 9 | 1465 | 53 | |
AY703043.1 | - | 8 - 9 | 2256 | 53 | |
AY703044.1 | - | 7 - 8 | 1950 | 53 | |
BC055285.1 | - | 9 - 10 | 2283 | 53 |