The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320032.1 | NP_001306961.1 | 5 - 6 | 776 | 86 |
NM_005688.3 | NP_005679.2 | 5 - 6 | 776 | 86 | |
XM_005247058.4 | XP_005247115.1 | 5 - 6 | 2344 | 86 | |
XM_005247059.4 | XP_005247116.1 | 5 - 6 | 766 | 86 | |
XM_011512314.1 | XP_011510616.1 | 5 - 6 | 667 | 86 | |
XM_011512315.1 | XP_011510617.1 | 5 - 6 | 826 | 86 | |
GenBank mRNA | AB019002.1 | - | 4 - 5 | 642 | 86 |
AB209103.1 | - | 2 - 3 | 286 | 86 | |
AB209454.1 | - | 5 - 6 | 703 | 86 | |
AF104942.1 | - | 5 - 6 | 715 | 86 | |
AF146074.1 | - | 5 - 6 | 786 | 86 | |
AY196484.1 | - | 4 - 5 | 602 | 86 | |
BC140771.1 | - | 4 - 5 | 652 | 86 | |
U83661.2 | - | 5 - 6 | 709 | 86 |