Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCCB
Entrez Gene ID
5096
Gene Name
propionyl-CoA carboxylase beta subunit
Gene Aliases
-
Chromosome Location
Chr.3: 136250325 - 136330171 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000532.4 NP_000523.2 5 - 6 590 101
NM_001178014.1 NP_001171485.1 6 - 7 650 101
XM_011512873.1 XP_011511175.1 5 - 6 590 101
GenBank mRNA AB209009.1 - 5 - 6 557 101
AF217984.1 - 2 - 3 496 101
AK130359.1 - 5 - 6 548 101
AK225215.1 - 5 - 6 545 101
AK225733.1 - 5 - 6 546 101
AK295312.1 - 6 - 7 617 101
AK302522.1 - 5 - 6 454 101
AL831978.2 - 5 - 6 540 101
BC013768.1 - 5 - 6 562 101
BC053661.1 - 5 - 6 557 101
S67325.1 - 5 - 6 569 101
X73424.1 - 5 - 6 609 101

View Full Product Details