Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEMA5B
Entrez Gene ID
54437
Gene Name
semaphorin 5B
Gene Aliases
SEMAG, SemG
Chromosome Location
Chr.3: 122909193 - 123028605 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031702.3 NP_001026872.2 10 - 11 1666 70
NM_001256346.1 NP_001243275.1 10 - 11 1806 70
NM_001256347.1 NP_001243276.1 10 - 11 1452 70
NM_001256348.1 NP_001243277.1 9 - 10 1504 70
NR_046079.1 10 - 11 1476 70
XM_017006638.1 XP_016862127.1 9 - 10 1716 70
XM_017006639.1 XP_016862128.1 9 - 10 1458 70
GenBank mRNA AB040878.1 - 10 - 11 1432 70
AK091481.1 - 10 - 11 1806 70
AK291407.1 - 10 - 11 1583 70
AK294528.1 - 9 - 10 1402 70
AK295619.1 - 10 - 11 1452 70
AK309848.1 - 9 - 10 1458 70
AY358124.1 - 10 - 11 1476 70
BC077726.1 - 10 - 11 1349 70

View Full Product Details