Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEMA5B
Entrez Gene ID
54437
Gene Name
semaphorin 5B
Gene Aliases
SEMAG, SemG
Chromosome Location
Chr.3: 122909193 - 123028605 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031702.3 NP_001026872.2 4 - 5 808 62
NM_001256346.1 NP_001243275.1 4 - 5 948 62
NM_001256347.1 NP_001243276.1 4 - 5 594 62
NM_001256348.1 NP_001243277.1 3 - 4 646 62
NR_046079.1 4 - 5 618 62
XM_017006638.1 XP_016862127.1 3 - 4 786 62
XM_017006639.1 XP_016862128.1 3 - 4 600 62
GenBank mRNA AB040878.1 - 4 - 5 574 62
AK091481.1 - 4 - 5 948 62
AK291407.1 - 4 - 5 725 62
AK294528.1 - 3 - 4 544 62
AK295619.1 - 4 - 5 594 62
AK309848.1 - 3 - 4 600 62
AY358124.1 - 4 - 5 618 62
BC077726.1 - 4 - 5 491 62

View Full Product Details