The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135095.1 | NP_001128567.1 | 16 - 17 | 1946 | 90 |
NM_022763.3 | NP_073600.3 | 16 - 17 | 2019 | 90 | |
XM_017007062.1 | XP_016862551.1 | 15 - 16 | 1717 | 90 | |
XM_017007063.1 | XP_016862552.1 | 11 - 12 | 1203 | 90 | |
XM_017007064.1 | XP_016862553.1 | 16 - 17 | 1999 | 90 | |
GenBank mRNA | AB098597.1 | - | 15 - 16 | 1904 | 90 |
AK075220.1 | - | 6 - 7 | 641 | 90 | |
AK127826.1 | - | 16 - 17 | 1999 | 90 | |
AK223599.1 | - | 16 - 17 | 2050 | 90 | |
AK314478.1 | - | 16 - 17 | 1946 | 90 | |
AY358146.1 | - | 9 - 10 | 892 | 90 | |
BC033635.2 | - | 4 - 5 | 494 | 90 | |
BC039297.1 | - | 16 - 17 | 1910 | 90 | |
BX648340.1 | - | 16 - 17 | 1941 | 90 |