Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FNDC3B
Entrez Gene ID
64778
Gene Name
fibronectin type III domain containing 3B
Gene Aliases
FAD104, PRO4979, YVTM2421
Chromosome Location
Chr.3: 172039628 - 172400703 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135095.1 NP_001128567.1 25 - 26 3401 75
NM_022763.3 NP_073600.3 25 - 26 3474 75
XM_017007062.1 XP_016862551.1 24 - 25 3172 75
XM_017007063.1 XP_016862552.1 20 - 21 2658 75
GenBank mRNA AB098597.1 - 24 - 25 3359 75
AF543840.1 - 7 - 8 1175 75
AK027052.1 - 9 - 10 1457 75
AK127826.1 - 25 - 26 3454 75
AK223599.1 - 25 - 26 3505 75
AK314478.1 - 25 - 26 3401 75
AL157482.1 - 7 - 8 1122 75
AY358146.1 - 18 - 19 2347 75
BC033635.2 - 13 - 14 1949 75
BC039297.1 - 25 - 26 3365 75
BX648340.1 - 25 - 26 3396 75

View Full Product Details