Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FNDC3B
Entrez Gene ID
64778
Gene Name
fibronectin type III domain containing 3B
Gene Aliases
FAD104, PRO4979, YVTM2421
Chromosome Location
Chr.3: 172039628 - 172400703 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135095.1 NP_001128567.1 8 - 9 1099 115
NM_022763.3 NP_073600.3 8 - 9 1172 115
XM_017007062.1 XP_016862551.1 7 - 8 870 115
XM_017007063.1 XP_016862552.1 3 - 4 356 115
XM_017007064.1 XP_016862553.1 8 - 9 1152 115
GenBank mRNA AB098597.1 - 7 - 8 1057 115
AK127826.1 - 8 - 9 1152 115
AK223599.1 - 8 - 9 1203 115
AK314478.1 - 8 - 9 1099 115
AY358146.1 - 1 - 2 45 115
AY358367.1 - 8 - 9 1141 115
BC039297.1 - 8 - 9 1063 115
BX648340.1 - 8 - 9 1094 115

View Full Product Details