Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RABGAP1L
Entrez Gene ID
9910
Gene Name
RAB GTPase activating protein 1 like
Gene Aliases
HHL, TBC1D18
Chromosome Location
Chr.1: 174159414 - 174995308 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AL157958.1 - 6 - 7 979 89

View Full Product Details