Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRG4
Entrez Gene ID
10216
Gene Name
proteoglycan 4
Gene Aliases
CACP, HAPO, JCAP, MSF, SZP
Chromosome Location
Chr.1: 186296273 - 186314562 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127708.2 NP_001121180.2 9 - 10 3726 112
NM_001127709.2 NP_001121181.2 8 - 9 3570 112
NM_001127710.2 NP_001121182.2 7 - 8 3447 112
NM_001303232.1 NP_001290161.1 9 - 10 3720 112
NM_005807.4 NP_005798.3 10 - 11 3849 112
XM_017000002.1 XP_016855491.1 7 - 8 3866 112
XM_017000003.1 XP_016855492.1 6 - 7 3433 112
GenBank mRNA AK057197.1 - 8 - 9 1685 112
AK131434.1 - 9 - 10 2420 112
U70136.1 - 9 - 10 3831 112

View Full Product Details