The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003292.2 | NP_003283.2 | 23 - 24 | 3389 | 117 |
GenBank mRNA | U69668.1 | - | 23 - 24 | 3180 | 117 |
X66397.1 | - | 23 - 24 | 3389 | 117 |