The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319960.1 | NP_001306889.1 | 3 - 4 | 563 | 71 |
NM_025191.3 | NP_079467.3 | 3 - 4 | 563 | 71 | |
NR_135118.1 | 3 - 4 | 563 | 71 | ||
XM_005245499.1 | XP_005245556.1 | 3 - 4 | 544 | 71 | |
XM_011510012.1 | XP_011508314.1 | 2 - 3 | 483 | 71 | |
XM_011510013.2 | XP_011508315.1 | 3 - 4 | 544 | 71 | |
XM_011510014.2 | XP_011508316.1 | 3 - 4 | 544 | 71 | |
XM_017002397.1 | XP_016857886.1 | 2 - 3 | 483 | 71 | |
XM_017002398.1 | XP_016857887.1 | 2 - 3 | 483 | 71 | |
GenBank mRNA | AF288393.1 | - | 3 - 4 | 226 | 71 |
AK225171.1 | - | 3 - 4 | 506 | 71 | |
AK315118.1 | - | 3 - 4 | 507 | 71 | |
BC105586.1 | - | 2 - 3 | 459 | 71 | |
BC144149.1 | - | 3 - 4 | 223 | 71 | |
BC144150.1 | - | 3 - 4 | 223 | 71 |