Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP1R12B
Entrez Gene ID
4660
Gene Name
protein phosphatase 1 regulatory subunit 12B
Gene Aliases
MYPT2, PP1bp55
Chromosome Location
Chr.1: 202348702 - 202588569 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197131.1 NP_001184060.1 6 - 7 502 69
NM_002481.3 NP_002472.2 23 - 24 3014 69
NM_032103.2 NP_115286.1 11 - 12 1369 69
XM_017001344.1 XP_016856833.1 24 - 25 3197 69
XM_017001345.1 XP_016856834.1 24 - 25 3194 69
XM_017001348.1 XP_016856837.1 23 - 24 3011 69
XM_017001350.1 XP_016856839.1 24 - 25 3071 69
XM_017001353.1 XP_016856842.1 14 - 15 1454 69
XM_017001355.1 XP_016856844.1 7 - 8 2198 69
GenBank mRNA AB003062.1 - 23 - 24 3027 69
AB050641.1 - 11 - 12 1369 69
AB588820.1 - 6 - 7 502 69
AF324888.1 - 23 - 24 3008 69
BC144699.1 - 23 - 24 3006 69

View Full Product Details