The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004947.4 | NP_004938.1 | 47 - 48 | 5028 | 59 |
XM_005264914.3 | XP_005264971.1 | 47 - 48 | 5300 | 59 | |
XM_005264915.3 | XP_005264972.1 | 47 - 48 | 5300 | 59 | |
XM_005264916.3 | XP_005264973.1 | 46 - 47 | 5051 | 59 | |
XM_005264917.3 | XP_005264974.1 | 47 - 48 | 5258 | 59 | |
XM_005264918.3 | XP_005264975.1 | 46 - 47 | 4930 | 59 | |
XM_006713009.3 | XP_006713072.1 | 46 - 47 | 5213 | 59 | |
XM_006713010.3 | XP_006713073.1 | 47 - 48 | 5063 | 59 | |
XM_011533441.2 | XP_011531743.1 | 36 - 37 | 5909 | 59 | |
XM_011533443.2 | XP_011531745.1 | 23 - 24 | 3590 | 59 | |
XM_011533444.2 | XP_011531746.1 | 23 - 24 | 3514 | 59 | |
XM_011533445.2 | XP_011531747.1 | 23 - 24 | 3535 | 59 | |
XM_017005825.1 | XP_016861314.1 | 47 - 48 | 5079 | 59 | |
XM_017005826.1 | XP_016861315.1 | 47 - 48 | 5069 | 59 | |
XM_017005827.1 | XP_016861316.1 | 38 - 39 | 4220 | 59 | |
XM_017005828.1 | XP_016861317.1 | 22 - 23 | 2945 | 59 | |
GenBank mRNA | AB002297.1 | - | 42 - 43 | 4636 | 59 |
AY145303.1 | - | 47 - 48 | 5005 | 59 | |
AY254099.1 | - | 47 - 48 | 5028 | 59 | |
BC131746.1 | - | 12 - 13 | 1396 | 59 |