Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEGF6
Entrez Gene ID
1953
Gene Name
multiple EGF like domains 6
Gene Aliases
EGFL3
Chromosome Location
Chr.1: 3487942 - 3624757 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001409.3 NP_001400.3 2 - 3 488 88
XM_011540885.1 XP_011539187.1 3 - 4 793 88
XM_011540886.1 XP_011539188.1 3 - 4 793 88
XM_011540888.2 XP_011539190.1 3 - 4 793 88
XM_017000533.1 XP_016856022.1 3 - 4 793 88
GenBank mRNA AB231860.1 - 2 - 3 488 88

View Full Product Details