The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080495.2 | NP_001073964.2 | 6 - 7 | 2602 | 56 |
XM_017012728.1 | XP_016868217.1 | 6 - 7 | 5099 | 56 | |
XM_017012729.1 | XP_016868218.1 | 5 - 6 | 2806 | 56 | |
XM_017012730.1 | XP_016868219.1 | 5 - 6 | 2424 | 56 | |
XM_017012731.1 | XP_016868220.1 | 5 - 6 | 2425 | 56 | |
XM_017012732.1 | XP_016868221.1 | 5 - 6 | 2425 | 56 | |
XM_017012733.1 | XP_016868222.1 | 4 - 5 | 2281 | 56 | |
XM_017012734.1 | XP_016868223.1 | 5 - 6 | 3307 | 56 | |
XM_017012735.1 | XP_016868224.1 | 5 - 6 | 2309 | 56 | |
XM_017012736.1 | XP_016868225.1 | 5 - 6 | 2896 | 56 | |
XM_017012737.1 | XP_016868226.1 | 5 - 6 | 4669 | 56 | |
XM_017012738.1 | XP_016868227.1 | 5 - 6 | 2191 | 56 | |
XM_017012739.1 | XP_016868228.1 | 5 - 6 | 2426 | 56 | |
XM_017012740.1 | XP_016868229.1 | 5 - 6 | 2426 | 56 | |
GenBank mRNA | CN295035.1 | - | 2 - 3 | 201 | 56 |
CR993399.1 | - | 2 - 3 | 459 | 56 |