Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYLIP
Entrez Gene ID
29116
Gene Name
myosin regulatory light chain interacting protein
Gene Aliases
IDOL, MIR
Chromosome Location
Chr.6: 16129086 - 16151015 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013262.3 NP_037394.2 6 - 7 1488 96
XM_005249032.2 XP_005249089.1 5 - 6 1323 96
XM_005249033.2 XP_005249090.1 5 - 6 1212 96
GenBank mRNA AF006003.1 - 6 - 7 1401 96
AF187016.2 - 6 - 7 1425 96
AF212221.1 - 5 - 6 1214 96
AF258586.1 - 6 - 7 1465 96
AK026739.1 - 3 - 4 707 96
AK074391.1 - 6 - 7 1488 96
AK314648.1 - 6 - 7 1488 96
BC002860.2 - 6 - 7 1469 96
BT007055.1 - 6 - 7 1251 96

View Full Product Details