The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030806.3 | NP_110433.1 | 1 - 2 | 360 | 71 |
GenBank mRNA | AF312864.1 | - | 1 - 2 | 282 | 71 |
AK056487.1 | - | 1 - 2 | 96 | 71 | |
AY258286.1 | - | 1 - 2 | 273 | 71 | |
BC028567.2 | - | 1 - 2 | 318 | 71 |