The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321153.1 | NP_001308082.1 | 17 - 18 | 2589 | 65 |
NM_007335.3 | NP_031361.2 | 17 - 18 | 2589 | 65 | |
NM_007337.3 | NP_031363.2 | 17 - 18 | 2589 | 65 | |
XM_006713438.3 | XP_006713501.1 | 17 - 18 | 3198 | 65 | |
XM_006713439.3 | XP_006713502.1 | 17 - 18 | 3197 | 65 | |
XM_006713440.3 | XP_006713503.1 | 17 - 18 | 3198 | 65 | |
XM_011534317.2 | XP_011532619.1 | 17 - 18 | 3197 | 65 | |
XM_011534318.2 | XP_011532620.1 | 16 - 17 | 3024 | 65 | |
GenBank mRNA | AB020522.1 | - | 17 - 18 | 2589 | 65 |
AB425198.1 | - | 17 - 18 | 2561 | 65 | |
BC129824.1 | - | 16 - 17 | 3405 | 65 | |
BC129825.1 | - | 17 - 18 | 2589 | 65 |