The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193374.1 | NP_001180303.1 | 2 - 3 | 152 | 74 |
NM_020415.3 | NP_065148.1 | 2 - 3 | 161 | 74 | |
GenBank mRNA | AF205952.1 | - | 1 - 2 | 139 | 74 |
AF290874.1 | - | 2 - 3 | 161 | 74 | |
AF323081.1 | - | 2 - 3 | 161 | 74 | |
AY207314.1 | - | 2 - 3 | 152 | 74 | |
AY359066.1 | - | 1 - 2 | 139 | 74 | |
BC069302.1 | - | 2 - 3 | 161 | 74 | |
BC101554.1 | - | 2 - 3 | 144 | 74 | |
BC101560.1 | - | 2 - 3 | 144 | 74 |