The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007208.3 | NP_009139.1 | 3 - 4 | 563 | 105 |
GenBank mRNA | AK296626.1 | - | 4 - 5 | 613 | 105 |
AK301377.1 | - | 3 - 4 | 397 | 105 | |
BC003375.2 | - | 3 - 4 | 462 | 105 | |
BI490082.1 | - | 3 - 4 | 532 | 105 | |
CR456720.1 | - | 3 - 4 | 368 | 105 | |
X06323.1 | - | 3 - 4 | 444 | 105 |