The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007208.3 | NP_009139.1 | 7 - 8 | 928 | 84 |
GenBank mRNA | AK296626.1 | - | 8 - 9 | 978 | 84 |
AK301377.1 | - | 7 - 8 | 762 | 84 | |
BC003375.2 | - | 7 - 8 | 827 | 84 | |
CR456720.1 | - | 7 - 8 | 733 | 84 | |
X06323.1 | - | 7 - 8 | 809 | 84 |