The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014188.2 | NP_054907.1 | 1 - 2 | 402 | 65 |
GenBank mRNA | AF161530.1 | - | 1 - 2 | 142 | 65 |
AF277178.1 | - | 1 - 2 | 81 | 65 | |
AJ276409.1 | - | 1 - 2 | 190 | 65 | |
AK001809.1 | - | 1 - 2 | 150 | 65 | |
AK023110.1 | - | 1 - 2 | 104 | 65 | |
AK291324.1 | - | 1 - 2 | 152 | 65 | |
AK297505.1 | - | 1 - 2 | 147 | 65 | |
AK310067.1 | - | 1 - 2 | 151 | 65 | |
BC008070.2 | - | 1 - 2 | 350 | 65 | |
BC018909.2 | - | 1 - 2 | 386 | 65 | |
CF594026.1 | - | - | 432 | 65 |