The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014188.2 | NP_054907.1 | 3 - 4 | 693 | 103 |
GenBank mRNA | AF161530.1 | - | 3 - 4 | 433 | 103 |
AF277178.1 | - | 3 - 4 | 372 | 103 | |
AJ276409.1 | - | 3 - 4 | 481 | 103 | |
AK001809.1 | - | 3 - 4 | 441 | 103 | |
AK127149.1 | - | 1 - 2 | 1629 | 103 | |
AK297505.1 | - | 3 - 4 | 438 | 103 | |
AK310067.1 | - | 3 - 4 | 442 | 103 | |
BC008070.2 | - | 3 - 4 | 641 | 103 | |
BC018909.2 | - | 3 - 4 | 677 | 103 |