Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ECD
Entrez Gene ID
11319
Gene Name
ecdysoneless cell cycle regulator
Gene Aliases
GCR2, HSGT1, SGT1
Chromosome Location
Chr.10: 73134524 - 73168095 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135752.1 NP_001129224.1 14 - 15 2046 105
NM_001135753.1 NP_001129225.1 12 - 13 1818 105
NM_007265.2 NP_009196.1 13 - 14 1947 105
NR_024203.1 11 - 12 1536 105
GenBank mRNA AK022616.1 - 13 - 14 1895 105
AK225519.1 - 12 - 13 1794 105
AK315711.1 - 14 - 15 2019 105
BC000721.2 - 13 - 14 1911 105
CR457407.1 - 12 - 13 1704 105
D88208.1 - 13 - 14 1911 105

View Full Product Details