The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077663.2 | NP_001071131.1 | 4 - 5 | 660 | 87 |
NM_001077664.2 | NP_001071132.1 | 4 - 5 | 432 | 87 | |
NM_001290075.1 | NP_001277004.1 | 4 - 5 | 655 | 87 | |
NM_001290076.1 | NP_001277005.1 | 5 - 6 | 554 | 87 | |
NM_017920.4 | NP_060390.3 | 3 - 4 | 633 | 87 | |
GenBank mRNA | AK314339.1 | - | 3 - 4 | 179 | 87 |
AL832134.1 | - | 2 - 3 | 2273 | 87 | |
AY078404.1 | - | 3 - 4 | 152 | 87 | |
BC018426.1 | - | 3 - 4 | 154 | 87 | |
BG722670.1 | - | 4 - 5 | 410 | 87 | |
BX640797.1 | - | 4 - 5 | 195 | 87 | |
BX648505.1 | - | 4 - 5 | 200 | 87 | |
CR749398.1 | - | 4 - 5 | 212 | 87 | |
DA240479.1 | - | 3 - 4 | 198 | 87 | |
DA316326.1 | - | 4 - 5 | 216 | 87 | |
DC336174.1 | - | 4 - 5 | 432 | 87 |