The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033017.3 | NP_148977.2 | 2 - 3 | 674 | 72 |
GenBank mRNA | AF220023.1 | - | 2 - 4 | 611 | 72 |