The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001096.2 | NP_001087.2 | 1 - 2 | 143 | 72 |
NM_198830.1 | NP_942127.1 | 1 - 2 | 143 | 72 | |
GenBank mRNA | AK304802.1 | - | 1 - 2 | 107 | 72 |
AK312315.1 | - | 1 - 2 | 147 | 73 | |
BC006195.2 | - | 1 - 2 | 111 | 72 | |
BG037168.1 | - | - | 163 | 72 | |
DB072009.1 | - | 1 - 2 | 147 | 73 | |
U18197.1 | - | 1 - 2 | 51 | 71 | |
X64330.1 | - | 1 - 2 | 64 | 71 |