The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001096.2 | NP_001087.2 | 20 - 21 | 2429 | 59 |
NM_001303274.1 | NP_001290203.1 | 20 - 21 | 2436 | 59 | |
NM_001303275.1 | NP_001290204.1 | 19 - 20 | 2406 | 59 | |
NM_198830.1 | NP_942127.1 | 19 - 20 | 2399 | 59 | |
XM_005257395.1 | XP_005257452.1 | 20 - 21 | 2478 | 59 | |
XM_017024688.1 | XP_016880177.1 | 19 - 20 | 2448 | 59 | |
GenBank mRNA | AB210035.1 | - | 20 - 21 | 2376 | 59 |
AK095084.1 | - | 10 - 11 | 1210 | 59 | |
AK295675.1 | - | 19 - 20 | 2406 | 59 | |
AK304802.1 | - | 12 - 13 | 1580 | 59 | |
AK312315.1 | - | 20 - 21 | 2433 | 59 | |
BC006195.2 | - | 20 - 21 | 2397 | 59 | |
U18197.1 | - | 20 - 21 | 2337 | 59 | |
X64330.1 | - | 20 - 21 | 2362 | 59 |