The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015613.2 | NP_056428.1 | 3 - 4 | 921 | 61 |
XM_011539623.2 | XP_011537925.1 | 2 - 3 | 1177 | 61 | |
XM_011539626.2 | XP_011537928.1 | 2 - 3 | 472 | 61 | |
GenBank mRNA | AB031547.1 | - | 3 - 4 | 61 |