The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270520.1 | NP_001257449.1 | 16 - 17 | 2223 | 66 |
NM_014992.2 | NP_055807.1 | 16 - 17 | 2147 | 66 | |
XM_005267430.1 | XP_005267487.1 | 17 - 18 | 2305 | 66 | |
XM_005267431.1 | XP_005267488.1 | 17 - 18 | 2405 | 66 | |
GenBank mRNA | AB014566.1 | - | 16 - 17 | 2133 | 66 |
AK021890.1 | - | 6 - 7 | 570 | 66 | |
AK291790.1 | - | 16 - 17 | 2192 | 66 | |
BC024781.1 | - | 6 - 7 | 864 | 66 | |
BC038428.1 | - | 16 - 17 | 2215 | 66 | |
BC064999.1 | - | 16 - 17 | 2206 | 66 |