The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004539.3 | NP_004530.1 | 3 - 4 | 477 | 126 |
XM_005266700.1 | XP_005266757.1 | 3 - 4 | 255 | 126 | |
GenBank mRNA | AJ000334.1 | - | 3 - 4 | 322 | 126 |
AK129897.1 | - | 3 - 4 | 280 | 126 | |
AK222835.1 | - | 3 - 4 | 324 | 126 | |
AK293985.1 | - | 2 - 3 | 257 | 126 | |
AK294364.1 | - | 3 - 4 | 324 | 126 | |
AK294763.1 | - | 3 - 4 | 308 | 126 | |
AK297777.1 | - | 3 - 4 | 317 | 126 | |
BC001687.1 | - | 3 - 4 | 326 | 126 | |
D84273.1 | - | 3 - 4 | 281 | 126 |