Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAD
Entrez Gene ID
790
Gene Name
carbamoyl-phosphate synthetase 2, aspartate transcarbamylase, and dihydroorotase
Gene Aliases
CDG1Z
Chromosome Location
Chr.2: 27217257 - 27243792 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306079.1 NP_001293008.1 17 - 18 2865 80
NM_004341.4 NP_004332.2 18 - 19 3054 80
XM_005264555.3 XP_005264612.1 18 - 19 3187 80
XM_006712101.2 XP_006712164.1 17 - 18 2998 80
GenBank mRNA AK226105.1 - 17 - 18 2847 80
BC065510.1 - 18 - 19 3054 80
D78586.1 - 18 - 19 2918 80

View Full Product Details