The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306079.1 | NP_001293008.1 | 21 - 22 | 3593 | 71 |
NM_004341.4 | NP_004332.2 | 22 - 23 | 3782 | 71 | |
XM_005264555.3 | XP_005264612.1 | 22 - 23 | 3915 | 71 | |
XM_006712101.2 | XP_006712164.1 | 21 - 22 | 3726 | 71 | |
GenBank mRNA | AK226105.1 | - | 21 - 22 | 3575 | 71 |
BC065510.1 | - | 22 - 23 | 3782 | 71 | |
BU541191.1 | - | 1 - 2 | 46 | 71 | |
D78586.1 | - | 22 - 23 | 3646 | 71 |