The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306079.1 | NP_001293008.1 | 36 - 37 | 5778 | 75 |
NM_004341.4 | NP_004332.2 | 37 - 38 | 5967 | 75 | |
XM_005264555.3 | XP_005264612.1 | 38 - 39 | 6151 | 75 | |
XM_006712101.2 | XP_006712164.1 | 37 - 38 | 5962 | 75 | |
GenBank mRNA | AK226105.1 | - | 36 - 37 | 5760 | 75 |
BC014178.2 | - | 12 - 13 | 1684 | 75 | |
BC065510.1 | - | 37 - 38 | 5967 | 75 | |
D78586.1 | - | 37 - 38 | 5831 | 75 |