The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304746.1 | NP_001291675.1 | 3 - 4 | 638 | 69 |
NM_017821.4 | NP_060291.2 | 3 - 4 | 668 | 69 | |
GenBank mRNA | AK097621.1 | - | 3 - 4 | 1403 | 69 |
AK294115.1 | - | 3 - 4 | 638 | 69 | |
AY126343.1 | - | 2 - 3 | 394 | 69 | |
BC137108.1 | - | 2 - 3 | 517 | 69 | |
BC137110.1 | - | 2 - 3 | 517 | 69 | |
BC144476.1 | - | 2 - 3 | 517 | 69 |