The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304746.1 | NP_001291675.1 | 5 - 6 | 852 | 119 |
NM_017821.4 | NP_060291.2 | 5 - 6 | 882 | 119 | |
GenBank mRNA | AK097621.1 | - | 5 - 6 | 1617 | 119 |
AK294115.1 | - | 5 - 6 | 852 | 119 | |
AY126343.1 | - | 4 - 5 | 608 | 119 | |
BC137108.1 | - | 4 - 5 | 731 | 119 | |
BC137110.1 | - | 4 - 5 | 731 | 119 | |
BC144476.1 | - | 4 - 5 | 731 | 119 |