The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000507.3 | NP_000498.2 | 6 - 7 | 1159 | 80 |
NM_001127628.1 | NP_001121100.1 | 7 - 8 | 1058 | 80 | |
XM_006717005.3 | XP_006717068.1 | 6 - 7 | 872 | 80 | |
GenBank mRNA | AF073475.1 | - | 6 - 7 | 825 | 80 |
AK292199.1 | - | 7 - 8 | 1097 | 80 | |
AK316583.1 | - | 6 - 7 | 1160 | 80 | |
AY544122.1 | - | 6 - 7 | 915 | 80 | |
BC012927.1 | - | 6 - 7 | 1025 | 80 | |
D26054.1 | - | 6 - 7 | 834 | 80 | |
D26055.1 | - | 6 - 7 | 834 | 80 | |
D26056.1 | - | 6 - 7 | 834 | 80 | |
L10320.1 | - | 6 - 7 | 970 | 80 | |
M19922.1 | - | 6 - 7 | 1026 | 80 |