The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166598.1 | NP_001160070.1 | 3 - 4 | 213 | 152 |
NM_052968.4 | NP_443200.2 | 3 - 4 | 237 | 152 | |
XM_017017169.1 | XP_016872658.1 | 2 - 3 | 695 | 152 | |
GenBank mRNA | AF202889.1 | - | 2 - 3 | 172 | 152 |
AF202890.1 | - | 2 - 3 | 172 | 152 | |
AK292739.1 | - | 2 - 3 | 184 | 152 | |
AK315229.1 | - | 2 - 3 | 184 | 152 | |
AY358749.1 | - | 2 - 3 | 175 | 152 | |
BC101787.1 | - | 2 - 3 | 200 | 152 | |
BC101789.1 | - | 2 - 3 | 200 | 152 | |
DA641693.1 | - | 3 - 4 | 213 | 152 | |
DB184692.1 | - | 2 - 3 | 208 | 152 |