The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000443.3 | NP_000434.1 | 20 - 21 | 2561 | 81 |
NM_018849.2 | NP_061337.1 | 20 - 21 | 2561 | 81 | |
NM_018850.2 | NP_061338.1 | 20 - 21 | 2561 | 81 | |
XM_011516308.2 | XP_011514610.2 | 20 - 21 | 2585 | 81 | |
XM_011516309.2 | XP_011514611.2 | 20 - 21 | 2585 | 81 | |
XM_011516310.2 | XP_011514612.2 | 19 - 20 | 2480 | 81 | |
XM_011516311.2 | XP_011514613.2 | 20 - 21 | 2585 | 81 | |
XM_011516312.2 | XP_011514614.2 | 20 - 21 | 2585 | 81 | |
XM_011516313.2 | XP_011514615.1 | 20 - 21 | 2584 | 81 | |
XM_011516315.2 | XP_011514617.2 | 15 - 16 | 1961 | 81 | |
XM_017012323.1 | XP_016867812.1 | 20 - 21 | 2572 | 81 | |
GenBank mRNA | BC042531.1 | - | 20 - 21 | 2597 | 81 |
M23234.1 | - | 20 - 21 | 2517 | 81 |