Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EIF2AK3
Entrez Gene ID
9451
Gene Name
eukaryotic translation initiation factor 2 alpha kinase 3
Gene Aliases
PEK, PERK, WRS
Chromosome Location
Chr.2: 88556740 - 88627576 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313915.1 NP_001300844.1 8 - 9 1298 158
NM_004836.6 NP_004827.4 8 - 9 1724 158
XM_017005376.1 XP_016860865.1 9 - 10 1616 158
GenBank mRNA AF110146.1 - 8 - 9 1491 158
AK128659.1 - 9 - 10 1477 158
AK308312.1 - 7 - 8 1092 158
AK315287.1 - 8 - 9 1629 158
BC126354.1 - 8 - 9 1518 158
BC126356.1 - 8 - 9 1515 158
CR749382.1 - 8 - 9 1299 158

View Full Product Details