The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024764.3 | NP_079040.2 | 19 - 20 | 2410 | 135 |
GenBank mRNA | AK024360.1 | - | 5 - 6 | 938 | 135 |
AK126034.1 | - | 19 - 20 | 2394 | 135 | |
BC126214.1 | - | 19 - 20 | 2364 | 135 | |
BC126216.1 | - | 19 - 20 | 2364 | 135 |