The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024764.3 | NP_079040.2 | 22 - 23 | 2867 | 83 |
GenBank mRNA | AK024360.1 | - | 8 - 9 | 1395 | 83 |
AK126034.1 | - | 22 - 23 | 2851 | 83 | |
BC126214.1 | - | 22 - 23 | 2821 | 83 | |
BC126216.1 | - | 22 - 23 | 2821 | 83 |