The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164232.1 | NP_001157704.1 | 17 - 18 | 2363 | 104 |
NM_015214.2 | NP_056029.2 | 17 - 18 | 2540 | 104 | |
XM_005273456.3 | XP_005273513.1 | 16 - 17 | 2598 | 104 | |
XM_017013252.1 | XP_016868741.1 | 17 - 18 | 2497 | 104 | |
XM_017013254.1 | XP_016868743.1 | 16 - 17 | 2277 | 104 | |
XM_017013255.1 | XP_016868744.1 | 15 - 16 | 2286 | 104 | |
GenBank mRNA | AB018268.1 | - | 14 - 15 | 1750 | 104 |
AK023218.1 | - | 13 - 14 | 1569 | 104 | |
AK026961.1 | - | 2 - 3 | 161 | 104 | |
AK056525.1 | - | 17 - 18 | 2442 | 104 | |
AK125904.1 | - | 17 - 18 | 2363 | 104 | |
AK127040.1 | - | 8 - 9 | 1891 | 104 | |
BC010504.1 | - | 12 - 13 | 1511 | 104 |