Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC16A14
Entrez Gene ID
151473
Gene Name
solute carrier family 16 member 14
Gene Aliases
MCT14
Chromosome Location
Chr.2: 230034974 - 230068999 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152527.4 NP_689740.2 2 - 3 724 87
XM_011510751.2 XP_011509053.1 2 - 3 495 87
XM_011510752.1 XP_011509054.1 2 - 3 724 87
XM_017003480.1 XP_016858969.1 2 - 3 418 87
GenBank mRNA AK123207.1 - 3 - 4 481 87
AK292873.1 - 2 - 3 456 87
BC034382.1 - 2 - 3 632 87
BC065524.1 - 2 - 3 628 87

View Full Product Details